Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Denatonium Benzoate 98.0+%, TCI America™
SDP

Catalog No. D2124100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2124100G 100 g
D21245G 5 g
D212425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2124100G Supplier TCI America Supplier No. D2124100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Molecular Weight (g/mol) 446.59
MDL Number MFCD00031578
InChI Key VWTINHYPRWEBQY-UHFFFAOYSA-N
Synonym denatonium benzoate, bitrex, lidocaine benzyl benzoate, anispray, aversion, gori, caswell no. 083bb, unii-m5ba6gaf1o, denatonium benzoate anhydrous
PubChem CID 19518
IUPAC Name benzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 100 g
Formula Weight 446.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Denatonium Benzoate
EINECSNumber (3)-2724
RTECSNumber BO6650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.