Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dendritic Cell Marker DCIR2 Monoclonal Antibody (33D1), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013271
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-132-71 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-132-71 Supplier Invitrogen™ Supplier No. 14588482
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer's patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii. Applications Reported: This 33D1 antibody has been reported for use in flow cytometric analysis, immunohistology staining of frozen tissue sections, and immunohistochemical staining. It has also been reported in complement-mediated cytotoxicity. Applications Tested: The 33D1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. For detection of purified 33D1 it is recommended not to use goat anti-rat due to its low affinity for 33D1. Rather, we recommend using biotin mouse anti-rat (cat. 13-4813).

The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer's patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dendritic Cell Marker DCIR2
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 33D1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Clec4a4
Gene Accession No. 0
Gene Alias Clec4a4; C-type lectin domain family 4, member a4; DC Marker; Dcir2; dendritic cell inhibitory receptor 2
Gene Symbols Clec4a4
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 474145
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.