Learn More
Creative Biomart Recombinant Dengue Type 1 Envelope Antigen, His-tagged
Supplier:  Creative Biomart DEN105
Encompass_Preferred
Recombinant Dengue Virus Subtype 1 Envelope 32 kDa containing domains I+II of the dengue envelope, produced in E. coli and fused with a 6 His tag.
Catalog No. 50-208-4483
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.