Learn More
Creative Biomart Recombinant Dengue Type 4 Envelope Antigen
Supplier:  Creative Biomart DEN118
Encompass_Preferred
The E. coli derived recombinant 28 kDa protein is genetically engineered peptide which is derived from Dengue Type-4 Envelope. This region also contains a common antigen for Dengue type 1, 2 and 3.
Catalog No. 50-208-4496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.