Learn More
Creative Biomart Recombinant Dengue Subtype 1 NS1 Antigen
Supplier:  Creative Biomart DNS127
Encompass_Preferred
Recombinant Dengue Virus NS1 Subtype 1 produced in insect cells is a polypeptide chain containing amino acids 777-1131 and having a molecular weight of approximately 50 kDa.Dengue NS1 ST1 is purified
Catalog No. 50-208-4505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.