Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral Dengue Virus 2 NS1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7113162
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB94393SP 25 μg
MAB94393100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB94393SP Supplier R&D Systems Supplier No. MAB94393SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus 2 NS1 Monoclonal specifically detects Dengue Virus 2 NS1 in Viral samples. It is validated for ELISA.

Specifications

Antigen Dengue virus 2 NS1
Applications ELISA
Classification Monoclonal
Clone 969822
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_739584
Gene Alias DEN-2, Dengue NS1, Dengue NS1 protein, Dengue Virus, Dengue virus Serotype 2, Dengue virus Type 2, DENV2, DENV2-NS1
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 2 NS1 protein Asp1-Ala352 Accession # NP_739584
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects recombinant Dengue Virus 2 NS1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.