Learn More
ZeptoMetrix Dengue Virus Type 1 (Strain: TC974) Culture Fluid
Shop All ZeptoMetrix Corporation Products

Description
- This product is intended for research, analytical testing during product development, or quality assurance testing. Viral culture fluids are sold as consumable testing materials, and are not for propagation or commercialization.
- Applications include: Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays.
- Quality Control
Specifications
Specifications
| Content And Storage | Viral culture fluids should be stored at -65°C or below. |
| Description | Repetitive freezing and thawing is not recommended (aliquot material if necessary). Titer will be altered by multiple freeze-thaws. |
| Product Type | Culture Fluid |
| For Use With (Application) | Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays |
| For Use With (Equipment) | To avoid cross-contamination, use separate pipette tips for all reagents. |
| Includes | Each frozen aliquot contains 1 mL of titered viral culture fluid. |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.