Learn More
Description
- Enveloped, icosahedral Flavivirus and contains a non-segmented, single stranded positive-sense RNA
- Propagated in the Vero cell line
- Purified using sucrose density gradient ultracentrifugation
- Disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO 13485 |
| Concentration | 0.5 to 3.0mg/mL |
| Content And Storage | -65°C or below |
| Detects | Viral antibodies |
| Form | Solution |
| Media Type | Vial |
| Product Type | Purified Viral Lysate |
| For Use With (Application) | Solid-phase enzyme immunoassays (EIA), western blot, dot blot and other protein-based assays |
| Purification Method | Sucrose density gradient ultracentrifugation |
| Show More |
For research use only. Not for use in diagnostic procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
