Learn More
Thermo Scientific™ Denka Seiken™ Agglutinating Sera E. coli Polyvalent
Description
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
- Types O29, O143, O152, O164
Specifications
Specifications
| Description | E. coli Polyvalent 7 |
| Detectable Analytes | E. coli |
| Type | Agglutinating Sera |
| Quantity | 2 mL/Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.