Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL151BJ Ladies Long Length Lab Coats
SDP

Catalog No. 22412149
Change view
Click to view available options
Color:
Ceil Blue
Green
Royal Blue
White
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-149 Royal Blue 4X-Large
22-412-115 White 2X-Small
22-412-119 White X-Small
22-412-116 Ceil Blue 2X-Small
22-412-117 Royal Blue 2X-Small
22-412-118 Green 2X-Small
22-412-120 Ceil Blue X-Small
22-412-121 Royal Blue X-Small
22-412-122 Green X-Small
22-412-124 Ceil Blue Small
22-412-126 Green Small
22-412-123 White Small
22-412-125 Royal Blue Small
22-412-127 White Medium
22-412-128 Ceil Blue Medium
22-412-129 Royal Blue Medium
22-412-135 White X-Large
22-412-137 Royal Blue X-Large
22-412-133 Royal Blue Large
22-412-131 White Large
22-412-130 Green Medium
22-412-134 Green Large
22-412-132 Ceil Blue Large
22-412-136 Ceil Blue X-Large
22-412-138 Green X-Large
22-412-139 White 2X-Large
22-412-142 Green 2X-Large
22-412-140 Ceil Blue 2X-Large
22-412-141 Royal Blue 2X-Large
22-412-145 Royal Blue 3X-Large
22-412-144 Ceil Blue 3X-Large
22-412-143 White 3X-Large
22-412-146 Green 3X-Large
22-412-148 Ceil Blue 4X-Large
22-412-150 Green 4X-Large
22-412-147 White 4X-Large
22-412-152 Ceil Blue 5X-Large
22-412-153 Royal Blue 5X-Large
22-412-151 White 5X-Large
22-412-154 Green 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
This item is not returnable. View return policy
Catalog No. 22-412-149 Supplier DenLine™ Uniforms Supplier No. DL151BJ4XLROY
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ladies full cut, a-line design lab coats are constructed with fluid and stain resistant fabric that meets AAMI PB70 Level 2 for use in BSL-1 and BSL-2 laboratories.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Dental
    • Human and Animal Research
  • Full cut design provides top of leg protection when seated
  • Complies with OSHA 29 CFR 1910, AAMI PB70 Level 2
  • Overall Air Permeability 18 CFM (ASTM D737)
  • Stain resistant fabric and cuffs (blood, bleach and all aqueous reagents)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Back belt detail
  • Hang loop outside of back of collar
  • Side vents for pant pocket access
  • Sizes run large and range from XXS to 5XL
  • Available White, Ceil Blue, Royal Blue and Green
  • 225 commercial wash tested, autoclavable fabric

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Women's
Material Polyester, Carbon
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Stains
Style Full-length
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Sleeve Style Tapered
Size (English) Hip 58 to 61.5 in.
Size (Metric) Hip 147 to 156 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental
Includes Hang Loop
Item Description With Back Belt Detail
Length (English) 39 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 42.5 in.
Length (Metric) 99 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.7 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 18 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.