Learn More
DenLine™ Uniforms Protection Plus™ DL151BJ Ladies Long Length Lab Coats

Description
- Primary Applications:
- Anatomic Pathology
- Clinical Pathology
- Clinical Laboratory
- Phlebotomy
- Specimen Collection
- Blood Collection, Testing
- Manufacturing
- Dental
- Human and Animal Research
- Full cut design provides top of leg protection when seated
- Complies with OSHA 29 CFR 1910, AAMI PB70 Level 2
- Overall Air Permeability 18 CFM (ASTM D737)
- Stain resistant fabric and cuffs (blood, bleach and all aqueous reagents)
- Anti-static fabric prevents false instrument readings, damage to computers
- Low lint fabric prevents sample contamination, ideal for molecular testing
- Durable construction, 3 pockets, double stitched
- Stainless snap closures
- Durable cuffs for overgloving, bleach safe all colors
- Back belt detail
- Hang loop outside of back of collar
- Side vents for pant pocket access
- Sizes run large and range from XXS to 5XL
- Available White, Ceil Blue, Royal Blue and Green
- 225 commercial wash tested, autoclavable fabric
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | 4X-Large |
| Gender | Women's |
| Material | Polyester, Carbon |
| Color | Green |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Fluids, Stains |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.