Learn More
DenLine™ Uniforms Protection Plus™ DL170BJ Unisex Long Length Lab Coats

Description
- Primary Applications:
- Anatomic pathology
- Clinical pathology and laboratory
- Phlebotomy
- Specimen and blood collection
- Manufacturing
- Animal research
- Cleanroom ISO 5, Class 100
- Highest barrier protection against strikethrough from incidental splash of blood
- Hydrostatic Pressure 32.5 CM (AATCC 127) and Impact Penetration 1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles
- Complies with OSHA 29 CFR 1910
- 360 Degree protection front as well as shoulders and back
- Fabric is fluid resistant, AAMI PB70 Level 2 Barrier
- High air permeability (35 CFM) is used to make the back of coat
- Overall air permeability of coat front and back is 18 CFM (ASTM D737)
- Stain resistant fabric and cuffs (blood, bleach and all aqueous reagents, liquids)
- Anti-static fabric prevents false instrument readings, damage to computers
- Low lint fabric prevents sample contamination, ideal for molecular testing
- Durable construction, 3 pockets, double-stitched, bar tacked
- Stainless snap closures
- Durable cuffs for overgloving, bleach safe all colors
- Hang loop, outside back of collar
- Side vents for pant pocket access
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | Polyester, Carbon |
| Color | Black |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Static, Fluids |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.