Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL170BJ Unisex Long Length Lab Coats
SDP

Catalog No. 22412187
Change view
Click to view available options
Color:
Black
Ceil Blue
Emerald Green
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-187 Black 4X-Large
22-412-006 Ceil Blue Small
22-412-003 Royal Blue X-Small
22-412-002 Ceil Blue X-Small
22-412-001 White X-Small
22-412-005 White Small
22-412-007 Royal Blue Small
22-412-009 White Medium
22-412-013 White Large
22-412-011 Royal Blue Medium
22-412-010 Ceil Blue Medium
22-412-014 Ceil Blue Large
22-412-015 Royal Blue Large
22-412-017 White X-Large
22-412-018 Ceil Blue X-Large
22-412-022 Ceil Blue 2X-Large
22-412-019 Royal Blue X-Large
22-412-021 White 2X-Large
22-412-023 Royal Blue 2X-Large
22-412-027 Royal Blue 3X-Large
22-412-029 White 4X-Large
22-412-026 Ceil Blue 3X-Large
22-412-030 Ceil Blue 4X-Large
22-412-025 White 3X-Large
22-412-031 Royal Blue 4X-Large
22-412-033 White 5X-Large
22-412-035 Royal Blue 5X-Large
22-412-034 Ceil Blue 5X-Large
22-412-180 Black X-Small
22-412-181 Black Small
22-412-182 Black Medium
22-412-183 Black Large
22-412-184 Black X-Large
22-412-186 Black 3X-Large
22-412-185 Black 2X-Large
22-412-188 Black 5X-Large
22-412-004 Emerald Green X-Small
22-412-008 Emerald Green Small
22-412-012 Emerald Green Medium
22-412-016 Emerald Green Large
22-412-020 Emerald Green X-Large
22-412-024 Emerald Green 2X-Large
22-412-028 Emerald Green 3X-Large
22-412-032 Emerald Green 4X-Large
22-412-036 Emerald Green 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
This item is not returnable. View return policy
Catalog No. 22-412-187 Supplier DenLine™ Uniforms Supplier No. DL170BJ4XLBLK
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wrinkle resistant, long length lab coats provide AAMI PB70 Level 2 fluid resistance and feature heavy duty stainless steel snap closures.

  • Primary Applications:
    • Anatomic pathology
    • Clinical pathology and laboratory
    • Phlebotomy
    • Specimen and blood collection
    • Manufacturing
    • Animal research
    • Cleanroom ISO 5, Class 100
  • Highest barrier protection against strikethrough from incidental splash of blood
  • Hydrostatic Pressure 32.5 CM (AATCC 127) and Impact Penetration 1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles
  • Complies with OSHA 29 CFR 1910
  • 360 Degree protection front as well as shoulders and back
  • Fabric is fluid resistant, AAMI PB70 Level 2 Barrier
  • High air permeability (35 CFM) is used to make the back of coat
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain resistant fabric and cuffs (blood, bleach and all aqueous reagents, liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double-stitched, bar tacked
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop, outside back of collar
  • Side vents for pant pocket access

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material Polyester, Carbon
Color Black
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static, Fluids
Style Long
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Size (English) Chest 58 to 60 in.
Size (English) Hip 54 to 57.5 in.
Size (Metric) Chest 147 to 152 cm
Size (Metric) Hip 137 to 146 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Length (English) 41 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 45.5 in.
Length (Metric) 104 cm
Length (Metric) Cuff 10 cm
Autoclavable Autoclavable
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.