Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine Protection Plus Unisex 41 in. (104 cm) Long Open Back Gown- Fluid Resistant Barrier
SDP

Catalog No. 22412100
Change view
Click to view available options
Color:
Ceil Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-100 Ceil Blue 3X-Large
22-412-089 White Small
22-412-093 White Large
22-412-091 White Medium
22-412-095 White X-Large
22-412-097 White 2X-Large
22-412-099 White 3X-Large
22-412-098 Ceil Blue 2X-Large
22-412-096 Ceil Blue X-Large
22-412-094 Ceil Blue Large
22-412-092 Ceil Blue Medium
22-412-090 Ceil Blue Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 22-412-100 Supplier DenLine Protection Plus Supplier No. DL1753XLCEI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Open Back Gown with One Piece Seamless Front and High Neck Coverage provides Wearer's Front, Shoulders Level 2 Protection from Incidental splash of Blood, Body Fluids, Aerosols, etc.; Open Back provides Exceptional Comfort to Wearer; Stain Resistant, Anti-static; 225 Commercial Wash Tested.

Primary Applications

· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics

Wearer Protection Provided

  • One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
  • Adjustable snaps at neck for complete coverage. Back ties at waist.
  • No patch pockets on front for splash of blood, body fluids, aerosols or OPIM to accumulate.
  • Open Back Design offers protection to shoulders from splash traveling either side of employee’s face.
  • AAMI PB70- Level 2, Assists wearers comply with OSHA 29 CFR 1910

Wearer Comfort

  • Overall Air Permeability of Front and Arms 12 CFM (ASTM D737)…Open Back
  • Raglan Sleeves and Loose Fit contribute to wearer comfort.

Other Features

  • Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
  • Anti-static Fabric prevents false instrument readings, damage to computers
  • Lint Free Fabric prevents sample contamination, ideal for molecular testing
  • Durable Construction
    • Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
    • Durable Cuffs for Overgloving – Bleach Safe all colors
  • Choice of 2 Colors, Ceil Blue and White; Sizes S to 3XL
  • 225 Commercial Wash Tested; Low Lifetime Cost per Wash
  • Autoclavable Fabric (274°F/135°C).
  • Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen

Specifications

Size 3X-Large
Material 99% Textured Polyester, 1% Carbon
Color Ceil Blue
Disposable Reusable
Closure Type Snap, Ties
Cuff Style Knit
Collar Type High
Finish Fluid and Stain Resistant, Anti-Static
Size (English) Chest 54 to 56 in.
Size (Metric) Chest 137 to 142 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
Length (Metric) 104.1 cm
Length (English) Cuff 4 in.
Length (Metric) Cuff 10.1 cm
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 12 CFM (ASTM D737)
Construction Fluid Resistant Front and Shoulders
Length (English) 41 in.
For Use With (Application) Protection Against Incidental Splash of Blood & OPIM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.