Learn More
DenLine™ Uniforms Protection Plus™ DL171 Unisex Mid-Length Lab Coats

Description
- Primary Applications:
- Anatomic Pathology
- Clinical Pathology
- Clinical Laboratory
- Phlebotomy
- Specimen Collection
- Blood Collection, Testing
- Manufacturing
- Human and Animal Research
- Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
- High barrier protection against strike through from incidental splash of blood
- Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles.
- Complies with OSHA 29 CFR 1910
- 360 degree protection front, shoulders, and back
- Fluid resistant fabric, AAMI PB70 Level 2 Barrier
- No cotton fabric used on shoulders or back as it would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head
- Mid-Length 34 in. coat provides wearers 5 ft. 4 in. and shorter frontal coverage to knees
- Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
- High air permeability (35 CFM) is used to make the back of coat for wearer comfort
- Overall air permeability of coat front and back is 18 CFM (ASTM D737)
- Stain fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
- Anti-static fabric prevents false instrument readings, damage to computers
- Low lint fabric prevents sample contamination, ideal for molecular testing
- Durable construction, 3 pockets, double stitched
- Stainless snap closures
- Durable cuffs for overgloving, bleach safe all colors
- Hang loop outside back of collar
- Side vents for pant pocket access
- Choice of 5 Colors White, Ceil Blue, Royal, Black and Green, Sizes XS to 3XL
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Polyester, Carbon |
| Color | Royal Blue |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Static, Fluids |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.