Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL171 Unisex Mid-Length Lab Coats
SDP

Catalog No. 22412062
Change view
Click to view available options
Color:
Black
Ceil Blue
Emerald Green
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
35 product options available for selection
Product selection table with 35 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-062 Ceil Blue 3X-Large
22-412-038 Ceil Blue X-Small
22-412-037 White X-Small
22-412-039 Royal Blue X-Small
22-412-041 White Small
22-412-042 Ceil Blue Small
22-412-047 Royal Blue Medium
22-412-045 White Medium
22-412-046 Ceil Blue Medium
22-412-043 Royal Blue Small
22-412-049 White Large
22-412-051 Royal Blue Large
22-412-054 Ceil Blue X-Large
22-412-050 Ceil Blue Large
22-412-053 White X-Large
22-412-055 Royal Blue X-Large
22-412-057 White 2X-Large
22-412-058 Ceil Blue 2X-Large
22-412-059 Royal Blue 2X-Large
22-412-061 White 3X-Large
22-412-063 Royal Blue 3X-Large
22-412-198 Black X-Small
22-412-199 Black Small
22-412-200 Black Medium
22-412-201 Black Large
22-412-203 Black 2X-Large
22-412-202 Black X-Large
22-412-204 Black 3X-Large
22-412-040 Emerald Green X-Small
22-412-044 Emerald Green Small
22-412-048 Emerald Green Medium
22-412-056 Emerald Green X-Large
22-412-052 Emerald Green Large
22-412-060 Emerald Green 2X-Large
22-412-064 Emerald Green 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 35 options available.
35 options
This item is not returnable. View return policy
Catalog No. 22-412-062 Supplier DenLine™ Uniforms Supplier No. DL1713XLCEI
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mid-length lab coats provide AAMI PB70 Level 2 fluid resistance against incidental splash of blood, body fluids, aerosols, and all aqueous liquids. Suitable for cleanroom use.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • High barrier protection against strike through from incidental splash of blood
  • Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles.
  • Complies with OSHA 29 CFR 1910
  • 360 degree protection front, shoulders, and back
  • Fluid resistant fabric, AAMI PB70 Level 2 Barrier
  • No cotton fabric used on shoulders or back as it would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head
  • Mid-Length 34 in. coat provides wearers 5 ft. 4 in. and shorter frontal coverage to knees
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • High air permeability (35 CFM) is used to make the back of coat for wearer comfort
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop outside back of collar
  • Side vents for pant pocket access
  • Choice of 5 Colors White, Ceil Blue, Royal, Black and Green, Sizes XS to 3XL

Specifications

Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material Polyester, Carbon
Color Ceil Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static, Fluids
Style Mid-length
No. of Pockets 3
Description Durable Knit Cuffs for Overgloving, Bleach Safe Colors
Collar Type Traditional
Latex Free Yes
Size (English) Chest 54 to 56 in.
Size (English) Hip 50 to 53.5 in.
Size (Metric) Chest 137 to 142 cm
Size (Metric) Hip 127 to 136 cm
Certifications/Compliance OSHA, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanroom ISO 5/Class 100
Length (English) 34 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 44 in.
Length (Metric) 86 cm
Length (Metric) Cuff 10.1 cm
Autoclavable Autoclavable
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.