Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DENND1A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983229
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3229 Supplier Bioss Supplier No. BS14269R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1A, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 (MIM 604199) and bind to clathrin and clathrin adaptor protein-2 (AP2). Thus, connecdenns link RAB35 activation with the clathrin machinery.

Specifications

Antigen DENND1A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DENND1A
Gene Accession No. Q8TEH3
Gene Alias 6030446I19Rik; BOS_12031; Connecdenn; Connecdenn 1; DENN domain containing 1A; DENN domain-containing protein 1A; DENN/MADD domain containing 1A; Dennd1a; FAM31A; KIAA1608; Protein FAM31A; RGD1307927; RP11-230L22.3
Gene Symbols DENND1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DENND1A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57706
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.