Learn More
Mettler Toledo™ Densito Charging Stand
Catalog No. 01915000 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Densito and DensitoPro Portable Density Meters
Catalog No. For Use With (Equipment)
01-915-000 Densito and DensitoPro Portable Density Meters
1 options
Catalog No. 01-915-000 Supplier Mettler Toledo™ Supplier No. 30330842
Add to Cart
Add to Cart
For use with Densito and DensitoPro Portable Density Meters
  • Charger for the lab and in production
  • With this stand, the Density2Go handheld meter is charged wirelessly
  • Configurable as stand or hang-up
  • The ideal accessory for viscous samples
  • When working with viscous samples, this stand stabilizes the Density2Go handheld meter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Accessory for Density Meter
For Use With (Equipment) Densito and DensitoPro Portable Density Meters