Learn More
United Scientific Supplies Density Cube Sets
SDPSCI_ED
Catalog No. S23979 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
10 Cubes
12 Cubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23979 10 Cubes
S23978 12 Cubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S23979 Supplier United Scientific Supplies Supplier No. DCSET10
Add to Cart
Add to Cart
Demonstrate a variety of densities with equal shaped cubes. The United Scientific Supplies Density Cube Sets include one of each of a variety of cubes for density studies.
  • For use in science experiments involving density
  • Cube measurements are equal
Specifications
Product Type Density Cubes
Description Wooden storage box
Includes One each of acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, and PVC cubes packaged in wooden storage box and a reference table
Quantity 10 Cubes
Height (English) 1 in.
Width (English) 1 in.