Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Density Kit
SDPSCI_ED

Catalog No. S81069 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Density Experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S81069 Density Experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81069 Supplier United Scientific Supplier No. DNSKITC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit is ideal for teaching students how to make measurements in the metric system, and then apply those measurements to determine density using the formula Density = Mass/Volume.

  • Ideal for teaching students how to make measurements in the metric system
  • Apply measurements to determine density using the formula Density = Mass/Volume
  • Includes metric ruler, aluminum bar (76 x 25 x 6 mm), aluminum cube (13 mm), steel sphere (13 mm) and glass sphere (13 mm)

Specifications

For Use With (Application) Density Experiments
Product Type Density Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.