Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Density of Liquids: The Color Column
GREENER_CHOICESCI_ED

Catalog No. S98652 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S98652 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol, which is known to the State of California to cause and reproductive harm.
Catalog No. S98652 Supplier Innovating Science™ Supplier No. IS7037
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol, which is known to the State of California to cause and reproductive harm.

Includes

Materials for five demonstrations: 450 mL glucose syrup, 400 mL liquid detergent, 400 mL vegetable oil, 400 mL ethyl alcohol, 25 mL blue food coloring, 25 mL red food coloring, graduated cylinder, plastic cups (5) and instructions

Witness a colorful density gradient.

  • Several liquids of varying color are added to a cylinder
  • A density gradient forms due to differing densities of the liquids, resulting in a layered, multi-colored column
  • As an optional investigation, compare the relative density of small solid objects by dropping them in the density column

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
Includes 450 mL glucose syrup, 400 mL liquid detergent, 400 mL vegetable oil, 400 mL ethyl alcohol un1170, 25 mL blue food coloring, 25 mL red food coloring, graduated cylinder, 5 plastic cups
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 1 Class
Product Type Density of Liquids: The Color Column Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.