Learn More
Eisco™ Density Metals Variety Set, 12 Pieces
SCI_ED
Catalog No. S27641 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S27641 1 Kit
1 options
Catalog No. S27641 Supplier Eisco™ Supplier No. PH0113
Add to Cart
Add to Cart
This 12 Piece Density Metals Variety Set provides learners a wide range of different values to study during density investigation, specific heat, and specific gravity exploration
  • Variety set includes an assortment of shapes and sizes
  • Includes 2 of each metal - Brass, Iron, Aluminum, Copper, Zinc & Lead
  • Masses range from 12 to 25 grams
  • Some oxidation or corrosion due to humidity and storage may occur due to the nature of the raw materials
  • Simply scuff any oxidation off to make the item look brand new again
  • Not fabricated using ultra high precision machining
  • Designed to be used in an educational setting
Specifications
Quantity 1 Kit
For Use With (Application) Density Investigation in Educational Settings
Product Type Density Metals Variety Set
Includes 2 of each: Brass, Iron, Aluminum, Copper, Zinc, Lead
Weight (Metric) 12 to 25 g
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov