Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Density Samples Sets Collection
SDPSCI_ED

Catalog No. S68357 Shop All Science First Products
Change view
Click to view available options
Density Samples Set Tekaform
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S68357 Density Samples Set Tekaform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68357 Supplier Science First Supplier No. 6112045

missing translation for 'validMainframeMsg'

Includes

  • Four sets: aluminum, brass, PVC and polypropylene
  • Wooden holder with linseed coating for water protection
  • Instructions

Each set of 12 is made from the same material in different lengths.

  • Determine the mass and volume of each sample to determine the density
  • Sets are available separately or as the complete collection

Specifications

For Use With (Application) To determine the mass and volume of each sample to determine the density
Product Type Density Sample Set
Includes Wooden holder with linseed coating for Water protection, instructions
Material Tekaform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.