Learn More
Sino Biological DENV DENV-E PAb 100UL 40471-T46-100
Supplier:  Sino Biological 40471T46100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein ( Catalog#40471-V08Y1; AAC59274.1; Met578-Gln680). Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein specific IgG was purified by Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein affinity chromatography.
Catalog No. 50-241-2159
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.