Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxycytidine-5'-Triphosphate, [α-32P], MP Biomedicals™

Catalog No. MP0133004X2 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 μCi
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0133004X2 250 μCi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0133004X2 Supplier MP Biomedicals, Inc Supplier No. 0133004X.2

Chemical Identifiers

Synonym dCTP a-32P

Specifications

Quantity 250 μCi
Formula Weight 871.9
Chemical Name or Material Deoxycytidine-5'-Triphosphate, [α-32P], Tetra-triethylammonium Salt
Recommended Storage Store at -20°C. Shipped in dry ice.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.