Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Deoxyribonuclease I, bovine pancreas, Molecular Biology Grade

Catalog No. AAJ60027EXU
Change view
Click to view available options
Quantity:
100 U
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60027EXU 500 U
AAJ60027EXR 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60027EXU Supplier Thermo Scientific Chemicals Supplier No. J60027EXU
Add to Cart

missing translation for 'validMainframeMsg'

Endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide and removal of DNA from protein samples. Also used in is used to nick DNA, as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used in the nuclease stock along with RNAse during cell lysate preparation of Madin-Darby canine kidney (MDCK) II cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 U
Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.