Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Deoxyribonucleic Acid
SCI_ED

Catalog No. S25294 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25294 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25294
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
Molecular Weight (g/mol) 0.00
MDL Number MFCD00130921
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
PubChem CID 44135672
IUPAC Name Deoxyribonucleic acid
SMILES *

Specifications

Assay Percent Range >100% w/v
Packaging Solid
Quantity 25 g
Chemical Name or Material Deoxyribonucleic Acid
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.