Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic acid, salmon testes

Catalog No. AAJ6084003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6084003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6084003 Supplier Thermo Scientific Chemicals Supplier No. J6084003
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9007-49-2 | C15H31N3O13P2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
Molecular Weight (g/mol) 0.00
MDL Number MFCD00130921
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
PubChem CID 44135672
IUPAC Name Deoxyribonucleic acid
SMILES *

Specifications

Packaging Plastic bottle
Quantity 1 g
Solubility Information Dissolve in TE buffer or water at 2mg/ml
Physical Form Lyophilized Powder
Chemical Name or Material Deoxyribonucleic acid
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.