Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic Acid Sodium Salt, MP Biomedicals™

Catalog No. MP216009805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP216009805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP216009805 Supplier MP Biomedicals, Inc Supplier No. 0216009805
Add to Cart
Add to Cart

Contains information in chemical code to direct the development of the cell according to its inheritance. Deoxyribonucleic Acid Sodium Salt, MP Biomedicals (DNA) is a polynucleotide and an essential component of chromosomes in cell nuclei. It is the carri

Deoxyribonucleic acid sodium salt has been used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin. Deoxyribonucleic acid sodium salt has also been used in a study to develop a simple method for extracting DNA from Cryptosporidium oocysts. It also acts as a template in the formation of ribonucleic acids, which play a fundamental role in the synthesis of proteins in the cell.

  • The purine and pyrimidine bases of the nucleosides are primarily adenine, guanine, cytosine and thymine
  • The sugar is D-2-deoxyribose
  • The nucleosides are linked together by phosphates in diester linkage from the 3'-hydroxyl of one sugar to the 5'-hydroxyl of the next
  • The repeating sugar-phosphate linkage forms the backbone of the single polynucleotide strand, which is the primary structure of DNA
  • Chemical analysis of DNA from different species show that the purine content is equal to the pyrimidine content; adenine content equal to thymine; guanine equal to cytosine
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
Molecular Weight (g/mol) 0.00
MDL Number MFCD00130921
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
PubChem CID 44135672
IUPAC Name Deoxyribonucleic acid
SMILES *

Specifications

Quantity 5 g
Solubility Information Most DNA is soluble in water or aqueous buffers. Slight heating may help. The addition of SDS or similar detergent may help solubilize DNA; however, detergents tend to denature the DNA into shorter strands.
Physical Form Powder
Recommended Storage 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.