Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic acid sodium salt, MP Biomedicals™

Catalog No. ICN16009801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16009801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN16009801 Supplier MP Biomedicals, Inc Supplier No. 0216009801

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
Molecular Weight (g/mol) 0.00
MDL Number MFCD00130921
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
PubChem CID 44135672
IUPAC Name Deoxyribonucleic acid
SMILES *

Specifications

Color White
Quantity 1 g
Physical Form Powder
Chemical Name or Material Deoxyribonucleic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.