Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic Acid Sodium Salt from Salmon or Herring Sperm , MP Biomedical

Catalog No. MP215227510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215227510 10 g
MP215227525 25 g
MP215227580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215227510 Supplier MP Biomedicals, Inc Supplier No. 0215227510
Add to Cart
Add to Cart

Deoxyribonucleic acid sodium salt has been used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin.

  • It is used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin.
  • It has also been used in a study to develop a simple method for extracting DNA from Cryptosporidium oocysts.
  • It contains information in chemical code to direct the development of the cell according to its inheritance.
  • The purine and pyrimidine bases of the nucleosides are primarily adenine, guanine, cytosine and thymine
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00283430
Synonym DNA Sodium Salt, DNA, Desoxiribon, Eucytol

Specifications

Color Cream
Loss on Drying ≤15%
Quantity 10 g
Solubility Information Most DNA is soluble in water or aqueous buffers. Slight heating may help. The addition of SDS or similar detergent may help solubilize DNA; however, detergents tend to denature the DNA into shorter strands.
Percent Purity 0.97
Physical Form Powder
RTECSNumber HG1933000
storageNote1 Store at +4°C. Store dessicated.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.