Learn More
Description
- 4-well panel containing antifungals incorporated into agar medium in different concentrations
- Terbinafine: 0.016 mg/L and 0.125 mg/L
- Itraconazole:1 mg/L
- Not for use directly with clinical or other specimens
- Microorganism to be tested must be pure culture on a dermatophyte culture medium supplemented with cycloheximide and chloramphenicol
- For mixed culture, selected colonies should be purified by subculturing
- Results are read visually at the end of incubation
Specifications
Specifications
| Form | Agar |
| Format | Dilution Panel |
| Manufacturing Origin | Italy |
| Product Type | Agar Medium |
| Sterility | Non-sterile |
| Target Organism Class | Fungi, Microconidia-forming dermatophytes |
| Test Type | Fungal Testing |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
