Learn More
Cell Biologics C57BL/6 Mouse Primary Dermal Lymphatic Endothelial Cells
SDP
Supplier:  Cell Biologics C576064L
Encompass_Preferred
C57BL/6 Mouse Primary Dermal Lymphatic Endothelial Cells can be used in assays of cell-cell adhesion, migration, vascular tube formation, or transendothelial resistance (TER). Standard biochemical procedures performed with endothelial cell cultures include RT-PCR, Western blotting, immunoprecipitation, immunofluorescent staining, immunofluorescent flow cytometry, or generating cell derivatives for desired research applications.
Catalog No. NC1631656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.