Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DES Mouse anti-Human, Clone: UMAB166, lyophilized, UltraMAB™
SDP

Catalog No. 501685036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5036 Supplier OriGene Supplier No. UM500107CF
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predomitly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.

Specifications

Antigen DES
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB166
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene DES
Gene Accession No. P17661
Gene Alias CDCD3, CMD1F, CSM1, CSM2, LGMD1D, LGMD1E, LGMD2R
Gene Symbols DES
Host Species Mouse
Immunogen Full length human recombit protein of human DES produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1674
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.