Learn More
CPC Scientific H-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ENKP004B
Encompass
SEQUENCE: H-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt)(Pen1 and Pen4 bridge)ONE-LETTER SEQUENCE: DPen-GF-Pen(Pen&Pen bridge)MOLECULAR FORMULA: C21H30N4O5S2MOLECULAR WEIGHT: 482.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:SKU(s): ENKP-004A, ENKP-004B
Catalog No. 50-210-9255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.