Learn More
Enzo Life Sciences DESC1 (human), (recombinant) (125U)
SDP
Supplier:  Enzo Life Sciences ALX2017511
Encompass_Preferred
Alternative name: Transmembrane protease serine 11E, TMPRSS11E. Concentration: 100nM. MW: ~45kDa; a-subunit: ~13kDa; b-subunit: ~32kDa. Formulation: Liquid. In 100 mM Tris-HCl, pH 8.0, containing 150 mM sodium chloride and 20% glycerol. Source: Produced in Drosophila Schneider 2 (S2) cells. Recombinant human DESC1 (aa 44-423) is fused at the C-terminus to a V5-His-tag. Produced by plasmid transfection expression. Long Term Storage: -80°C. Use/Stability: Stable for at least 6 months when stored at -20°C. Recommended Assay Buffer: 100mM TRIS, pH 9.0, containing 5mg/ml BSA. Do not store diluted solutions. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7980
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.