Learn More
Cayman Chemical DescarbamylnovobIocIn 5mg
SDP
Supplier:  Cayman Chemical 299715 MG
Encompass_Preferred
A selective adenosine A1 receptor agonist (Kis 3 1 and 1300 nM for adenosine A1 and A2 receptors respectively in rat brain inhibits isoprenaline-induced cAMP accumulation in DDT1MF-2 hamster smooth muscle cells (IC50 2 6 nM) but only decreases contractility in isolated guinea pig atria with an EC50 value of 1500 nM inhibits norepinephrine-induced lipolysis in rat adipocytes (EC50 0 83 nM) decreases locomotor activity and inhibits DMCM-induced seizures in mice (ED50s 0 13 and 0 3 mg/kg respectively) decreases blood pressure by 74 in anesthetized rats at 0 1 mg/kg
Catalog No. 50-298-8657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.