Learn More
Desiccant Packs, Cytiva
Catalog No. 09928142 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 Pk./Cs.
Catalog No. Quantity
09-928-142 100 Pk./Cs.
1 options
Catalog No. 09-928-142 Supplier Cytiva Supplier No. 10548239
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Edge
Add to Cart
Ensure that DNA sampling cards remain dry during transportation or storage
  • High vapor transmission rate
  • Substrate made with Tyvek™ material
  • Non-dusting, strong and hard to tear
TRUSTED_SUSTAINABILITY
Specifications
Material Tyvek™ Substrate
Quantity 100 Pk./Cs.
Weight 1 g