Learn More
Hach Company Desiccator, Vacuum, 171 mm (6.7") diameter
Supplier: Hach Company 2238300
Round polycarbonate top provides lightweight vessel with unmatched transparency. Polypropylene lower section creates support chamber at full vacuum. Stopcock for vacuum applications (1/4-3/8" OD vacuum tubing, sold separately). Holds 28 in Hg (711 mm) for 24 hrs. Silicone O-ring makes grease unnecessary. For room temperature, non-organic solvent, and non-autoclave uses only.