Learn More
Cayman Chemical Desmethyl SIbutramInhydroc 1mg
SDP
Supplier:  Cayman Chemical 350731 MG
Encompass_Preferred
A peptide with diverse biological activities an agonist of the GRPR and the NMB receptor in BALB/c 3T3 fibroblasts expressing the human receptors (EC50s 0 44 and 0 03 nM respectively) binds to frog BB4 in CHO-K1 cells (Ki 1 2 nM) induces contractions of isolated guinea pig gallbladder at 10-100 ng/kg decreases food intake in rats at 4-128 Ug/kg
Catalog No. 50-291-0480
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.