Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoglein 1 Monoclonal Antibody (27B2)
GREENER_CHOICE

Catalog No. 326000
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-600-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-600-0 Supplier Invitrogen™ Supplier No. 326000
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Desmoglein 1 Monoclonal Antibody for Western Blot, IHC (P), IHC (F), IP

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein 1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 27B2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene DSG1
Gene Accession No. Q02413
Gene Alias Cadherin family member 4; CDHF4; desmoglein 1; desmoglein 1 beta; desmoglein 1 gamma; desmoglein-1; Desmosomal glycoprotein 1; DG1; DSG; DSG1; Dsg1b; Dsg1c; EPKHE; EPKHIA; Pemphigus foliaceus antigen; PPKS1; SPPK1
Gene Symbols DSG1
Host Species Mouse
Immunogen Human desmoglein-1 intracellular domain.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1828
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.