Learn More
Invitrogen™ Desmoplakin Monoclonal Antibody (DP-2.15)

Description
Mouse anti Desmoplakin 1/2 antibody, clone DP-2.15 recognizes both desmoplakin 1 and 2 from stratified epithelia, simple epithelia including glands, urothelium, thymic reticular epithelium, hepatocytes, intercalated disks of myocardium and arachnoid cells of meninges. Mouse anti Desmoplakin 1/2 antibody, clone DP-2.15 can be used for the detection of primary and metastatic carcinomas.
Specifications
Specifications
| Antigen | Desmoplakin |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | DP-2.15 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | DSP |
| Gene Accession No. | E9Q557, P15924 |
| Gene Alias | 2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.