Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Destrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588166 Supplier Invitrogen™ Supplier No. PA588166
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASGVQVADE VCRIFYDMKV RKCSTPEEIK KRKKAVIFCL SADKKCIIVE EGKEILVGDV GVTITDPFKH FVGMLPEKDC RYALYDASFE TKESRKEELM FFLWAPELAP LKSKMIYASS KDAIKKKFQG IKHECQANGP EDLNRACIAE KLGGSLIVAF EGCPV; Positive Samples: A-549, HeLa.

The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Destrin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DSTN
Gene Accession No. P60981
Gene Alias 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p
Gene Symbols DSTN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human Destrin (NP_0068611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11034
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.