Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desvenlafaxine, 97%

Catalog No. AC461570010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461570010 1 g
AC461570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461570010 Supplier Thermo Scientific Chemicals Supplier No. 461570010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 93413-62-8 | C16H25NO2 | 263.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93413-62-8
Molecular Formula C16H25NO2
Molecular Weight (g/mol) 263.38
InChI Key KYYIDSXMWOZKMP-UHFFFAOYSA-N
Synonym desvenlafaxine, o-desmethylvenlafaxine, 4-2-dimethylamino-1-1-hydroxycyclohexyl ethyl phenol, pristiq, o-desmethyl venlafaxine, desvenlafaxine inn:ban, desvenlafaxine inn, o-desmethylvenlafaxine odv, 1-2-dimethylamino-1-4-hydroxyphenyl ethyl cyclohexanol
PubChem CID 125017
ChEBI CHEBI:83527
IUPAC Name 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
SMILES CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 263.38
Percent Purity 97%
Chemical Name or Material Desvenlafaxine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.