Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detecting COVID-19 Using Reverse-Transcription PCR (RT-PCR)
SDPSCI_ED

Catalog No. S52005 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students (5 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S52005 25 Students (5 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52005 Supplier Edvotek™ Supplier No. 363
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this simulated RT-PCR experiment, students will explore the diagnostic test used worldwide to diagnose and monitor the spread of COVID-19.

  • This experiment requires a PCR thermal cycler
  • Please note, this experiment is a DNA simulation of reverse transcription PCR and is intended solely for educational purposes. There are no viral or human DNA products included in this experiment. The experiment will not perform viral RNA testing for SARS-CoV-2
  • For 25 students working in 5 groups
  • Time required: Set up - 30 minutes, PCR - 2 hours or overnight, Electrophoresis - 45 minutes
  • Instructions, PCR EdvoBeads™, LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, DNA Templates, TE Buffer, UltraSpec-Agarose™, TBE Electrophoresis Buffer powder, SYBR™ Safe Stain, FlashBlue™ DNA Stain, Microcentrifuge & PCR Tubes
  • Some components require freezer storage upon receipt

Specifications

Grade 9 to Undergraduate
Age 15 to 20
For Use With (Application) Explore the diagnostic test used worldwide to diagnose and monitor the spread of COVID-19.
Includes Instructions, PCR EdvoBeadsTM, LyphoPrimerTM Mix, EdvoQuickTM DNA Ladder, DNA Templates, TE Buffer, UltraSpec-AgaroseTM, TBE Electrophoresis Buffer powder, SYBR™ Safe Stain, FlashBlueTM DNA Stain, Microcentrifuge & PCR Tubes
Requires DNA Electrophoresis, Thermal Cycler, Micropipettes: 5-50 μL, UV or Blue Light Transilluminator, White Light Box (optional - use if staining with FlashBlueTM), Microcentrifuge & Microwave or Hot plate.
Content And Storage Some Components Require Freezer Storage Upon Receipt
Class Size 25 Students (5 Groups)
Product Type PCR Kit

**Please note, this experiment is a DNA simulation of reverse transcription PCR and is intended solely for educational purposes. There are no viral or human DNA products included in this experiment. The experiment will not perform viral RNA testing for SARS-CoV-2.**

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.