Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Detergent, Neutral Reagent, Fisher Chemical™

Catalog No. SF11820 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF118-20 20 L PolyPac™
SF118-4 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF118-20

Components: Triethylene Glycol (<1.5%), SDS (3.0%), Borates (each <1.0%), EDTA (<2.0%), and Water (to balance)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Detergent, Neutral Reagent
Color Colorless
Physical Form Liquid
Boiling Point 100°C
Melting Point 0°C
CAS 151-21-3, 6381-92-6, 112-27-6, 1303-96-4
Quantity 20 L
Identification Pass Test
Packaging PolyPac™
MDL Number MFCD00283948
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.