Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Detergent Removal Spin Columns & Plates

Catalog No. PI87778
Change view
Click to view available options
Quantity:
10 mL
2 x 96-well Plates
24 x 0.1 mL Columns
25 x 125 μL Columns
25 x 500 μL Columns
5 mL
5 x 2 mL Columns
5 x 4 mL Columns
Product Line:
HiPPR
Pierce
Product Type:
Detergent Removal Resin
Spin Column
Spin Plate
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Line Product Type
PI87778 5 x 2 mL Columns Pierce Spin Column
PI87780 10 mL Pierce Detergent Removal Resin
PI87776 25 x 125 μL Columns Pierce Spin Column
PI87777 25 x 500 μL Columns Pierce Spin Column
PI87779 5 x 4 mL Columns Pierce Spin Column
PI88304 2 x 96-well Plates Pierce Spin Plate
PI88305 5 mL HiPPR Spin Column
PI88306 24 x 0.1 mL Columns HiPPR Spin Column
PI88307 2 x 96-well Plates HiPPR Spin Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. PI87778 Supplier Thermo Scientific™ Supplier No. 87778
Add to Cart
Add to Cart

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.

Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.

  • Optimized—removes >95% of detergent from samples of proteins or peptides
  • Fast—sample processing takes less than 15 minutes
  • Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin

Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit

HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:

  • Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
  • Spin columns—0.1 mL micro-centrifuge columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification
TRUSTED_SUSTAINABILITY

Specifications

Description 15 mL Conical, Spin Column
Product Type Spin Column
Content And Storage Upon receipt store at 4°C. The resin is stored in 0.15 M NaCl, 0.05% sodium azide.
Column Type Proprietary Resin, Affinity
Stationary Phase Proprietary Ligand
Product Line Pierce
Purification Target Detergent Removal
Sufficient For 150 to 500 μL Sample Each Column
Quantity 5 x 2 mL Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.