Learn More
G-Biosciences DetergentOUT™ Tween™, Gravity Flow Column
SCI_ED
Catalog No. S000782
Change view
Click to view available options
Quantity:
10 x 15 mL
10 x 30 mL
1 x 150 mL
5 x 150 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000782 10 x 15 mL
S000814 10 x 30 mL
S000815 1 x 150 mL
S000797 5 x 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S000782 Supplier G-Biosciences Supplier No. 7862021
Add to Cart
Add to Cart
Pre-packed, spin and gravity flow column format, detergent removal resin columns for polysorbate or Tween™ detergents or surfactants.
The resin in the columns has a high binding affinity for polysorbate detergents, commercially known as Tween™. The binding capacity is approx 85 mg polysorbate detergent per 1 mL resin. The resin does not bind to protein allowing for a >95% protein recovery.
Specifications
Product Type Detergent Removal Resin
Form Solution
Product Line Pierce
Quantity 10 x 15 mL