Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Determination of Blood Type Using Real Blood and Saliva Kit
GREENER_CHOICESCI_ED

Catalog No. S20637 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S20637 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20637 Supplier Innovating Science™ Supplier No. IS3190

Designed to allow students to test their own saliva to determine their ABO blood type.

  • Initial box contains all of the non-perishable supplies including a disinfecting solution spray
  • Allows the students to better understand how real blood typing works and what would happen if the wrong blood was given to a patient needing a transfusion
  • Titers will be performed using supplied screened/typed human blood samples
  • Students that are part of the ∼80% of the human population that secrete their blood antigens will perform titers to understand which blood type they belong to

Kit Includes:

  • 99.9% Isopropyl Alcohol (500 mL)
  • 0.9% Saline (150 mL)
  • 30 Saliva Collection Cups
  • 150 Graduated

Coupon for perishable materials:
Redeem by fax, phone or e-mail. Materials sent upon redemption of coupon

  • Anti A (10 mL)
  • Anti B (10 mL)
  • Red Blood Cells Type A (10 mL)
  • Red Blood Cells Type B (10 mL)
  • Red Blood Cells Type O (10 mL)
  • H Lectin (10 mL)

Specifications

Grade 7 to 14
Description For 30 students working in groups of 3
Format Kit
Product Type Determination of Blood Type Using Real Blood and Saliva
Includes Redeemable coupon for perishable materials
Quantity 1 Kit
Color White
Languages English
For Use With (Application) Determining ABO blood types
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.