Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Deubiquitinase Assay Kit, Fluorescent, Cell and Tissue Extracts, 100 Tests

Catalog No. 502586026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6026 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6026 Supplier Abcam Supplier No. AB241002100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent enzyme activity assay for measuring deubiquitinase in cell and tissue extracts.

  • Applications: Enzyme activity assay
  • Detection method: Fluorescent
  • Sample types: Cell culture extracts, Tissue extracts
  • Equipment compatibility: Microplate reader
  • Citation count: 3

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Deubiquitinase
No. of Reactions 100 Tests
Detection Location In-Solution Detection
For Use With (Equipment) Microplate Reader
Detection Method Fluorescence
Format Kit
Sample Type Cell Culture Extracts, Tissue extracts
Quantity 100 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.